missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NOD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566325
Description
Immunogen sequence: QHLRVLHLPFS HLGPGGALSL AQALDGSPHL EEISLAENNL AGGVLRFCME LPLLRQIDLV SCKIDNQTAK LLTSSFTSCP ALEVILLSWN LLGDE Highest antigen sequence identity to the following orthologs - mouse 68%, rat 65%.
NOD4 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD4 contains a CARD-like domain, a central NOD domain and a large LRR region. NOD4, an IFN-gamma-inducible nuclear protein, plays a role in homeostatic control of innate immunity and in antiviral defense mechanisms. As a key negative regulator of NF-kappa-B and type I interferon signaling, NOD4 may be a useful target for manipulating immune responses against infectious or inflammation-associated diseases, including cancer.Specifications
| NOD4 | |
| Polyclonal | |
| Unconjugated | |
| NLRC5 | |
| AI451557; AK220210; Caterpiller protein 16.1; CLR16.1; copine II; Cpne2; NLR family CARD domain containing 5; NLR family, CARD domain containing 5; Nlrc5; NOD27; NOD4; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; protein NLRC5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WI3 | |
| NLRC5 | |
| Recombinant Human NOD4. Recombinant protein control fragment (Product #RP-107006). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |