missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOC2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575404 Supplier Invitrogen™ Supplier No. PA575404
Only null left

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

INHAT (inhibitor of acetyltransferases) complex binds to histones and masks them from being substrate for acetyltransferase. Endogenous INHAT subunits, which include the Set/TAF-Ibeta oncoprotein, associate with chromatin in vivo and can block coactivator mediated transcription when transfected in cells.

Specifications

Antigen NOC2L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NOC2L
Gene Accession No. Q9WV70, Q9Y3T9
Gene Alias AA410003; AF155546; DKFZp564C186; FLJ35172; hypothetical protein LOC508638; NET15; NET7; NIR; NOC2 like nucleolar associated transcriptional repressor; NOC2L; NOC2L nucleolar associated transcriptional repressor; NOC2-like; NOC2-like nucleolar associated transcriptional repressor; NOC2-like protein; novel INHAT (inhibitor of histone acetyltransferase) repressor; novel INHAT repressor; nucleolar complex associated 2 homolog; nucleolar complex associated 2 homolog (S. cerevisiae); nucleolar complex protein 2 homolog; PPP1R112; Protein NOC2 homolog; protein phosphatase 1, regulatory subunit 12; RGD1309387
Gene Symbols NOC2L
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 602-650 of Human NOC2L.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26155, 313777, 57741
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less