missing translation for 'onlineSavingsMsg'
Learn More

Nobiletin 95.0+%, TCI America™

$294.52 - $1614.27

Chemical Identifiers

CAS 478-01-3
Molecular Formula C21H22O8
Molecular Weight (g/mol) 402.399
MDL Number MFCD03273560
InChI Key MRIAQLRQZPPODS-UHFFFAOYSA-N
PubChem CID 72344
ChEBI CHEBI:7602
IUPAC Name 2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C(=C3OC)OC)OC)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N087110MG
View Documents
TCI America
N087110MG
10 mg
Each for $294.52
Only null left
 
N0871100MG
View Documents
TCI America
N0871100MG
100 mg
Each for $1,614.27
Only null left
 
Description

Description

Specifications

Chemical Identifiers

478-01-3
402.399
MRIAQLRQZPPODS-UHFFFAOYSA-N
CHEBI:7602
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C(=C3OC)OC)OC)OC)OC
C21H22O8
MFCD03273560
72344
2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one

Specifications

478-01-3
White-Yellow
MFCD03273560
3′C,4′C,5,6,7,8-Hexamethoxyflavone
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C(=C3OC)OC)OC)OC)OC
402.399
CHEBI:7602
≥95.0% (HPLC)
Nobiletin
138°C
C21H22O8
10 mg
MRIAQLRQZPPODS-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one
72344
402.40
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ3052200

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'