missing translation for 'onlineSavingsMsg'
Learn More

Nobiletin 95.0+%, TCI America™

Supplier:  TCI America N087110MG

 View more versions of this product

Catalog No. N087110MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nobiletin
138°C
C21H22O8
10 mg
MRIAQLRQZPPODS-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one
72344
402.40
Crystalline Powder
478-01-3
White-Yellow
MFCD03273560
3′C,4′C,5,6,7,8-Hexamethoxyflavone
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C(=C3OC)OC)OC)OC)OC
402.399
CHEBI:7602
≥95.0% (HPLC)

Chemical Identifiers

478-01-3
402.399
MRIAQLRQZPPODS-UHFFFAOYSA-N
CHEBI:7602
COC1=C(C=C(C=C1)C2=CC(=O)C3=C(O2)C(=C(C(=C3OC)OC)OC)OC)OC
C21H22O8
MFCD03273560
72344
2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxychromen-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ3052200

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions