missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NMNAT1 (Human) Recombinant Protein
Human NMNAT1 full-length ORF ( NP_073624.2, 1 a.a. - 279 a.a.) recombinant protein with GST-tag at N-terminal.
Supplier: Abnova™ H00064802P012
Description
- Sequence: MENSEKTEVVLLACGSFNPITNMHLRLFELAKDYMNGTGRYTVVKGIISPVGDAYKKKGLIPAYHRVIMAELATKNSKWVEVDTWESLQKEWKETLKVLRHHQEKLEASDCDHQQNSPTLERPGRKRKWTETQDSSQKKSLEPKTKAVPKVKLLCGADLLESFAVPNLWKSEDITQIVANYGLICVTRAGNDAQKFIYESDVLWKHRSNIHVVNEWIANDISSTKIRRALRRGQSIRYLVPDLVQEYIEKHNLYSSESEDRNAGVILAPLQRNTAEAKT
Specifications
| NP_073624.2 | |
| nicotinamide nucleotide adenylyltransferase 1 | |
| 125% SDS-PAGE Stained with Coomassie Blue | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| NMNAT1 | |
| GST |
| 64802 | |
| Wheat germ expression system | |
| 2 μg | |
| NMNAT, PNAT-1, PNAT1 | |
| Wheat Germ (in vitro) | |
| 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |