Learn More
Invitrogen™ NLGN4X (Neuroligin-4) Monoclonal Antibody (CSTEM30), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 14915880
Description
This monoclonal antibody CSTEM30 reacts with human Neuroligin-4, X linked, also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X. Applications Reported: This CSTEM30 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. (Fluorochrome-conjugated CSTEM30 is recommended for use in flow cytometery.) Applications Tested: This CSTEM30 antibody has been tested by immunocytochemistry on formaldehyde-fixed and permeabilized human iPSCs, and by flow cytometry on human iPSCs. For immunocytochemistry, this can be used at less than or equal to 5 μg/mL. For flow cytometry, this antibody can be used at less than or equal to 0.5 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Neuroligin-4, X linked (also known as NLGN4X, NLGN4, Neuroligin-4, and Neuroligin 4X) is a postsynaptic cell-adhesion molecule that interacts with presynaptic cell-surface neurexins. These interactions bridge together the synaptic cleft and have an essential role in synaptic transmissions between neurons. There are 5 neuroligin genes in humans: NLGN1, NLGN2, NLGN3, NLGN4X, and NLGN4Y. Mutations in NLGN3 and NLGN4X are linked to Autism Spectrum Disorders (ASD). The expression of neuroligin-4, X-linked has been reported on human pluripotent stem cells.Specifications
| NLGN4X (Neuroligin-4) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with proprietary stabilizer and 0.09% sodium azide; pH 7.2 | |
| Q8N0W4 | |
| NLGN4X | |
| Affinity chromatography | |
| RUO | |
| 57502 | |
| 4°C, do not freeze | |
| Liquid |
| Flow Cytometry, Immunocytochemistry | |
| CSTEM30 | |
| Unconjugated | |
| NLGN4X | |
| ASPGX2; AUTSX2; HLNX; HNL4X; HNLX; KIAA1260; MGC22376; neuroligin 4, X-linked; neuroligin 4-like; Neuroligin X; Neuroligin-4, X-linked; NL4; NL-4; NLGN; Nlgn4; Nlgn4l; NLGN4X; UNQ365/PRO701 | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |