missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NKRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5150276
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5150276 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5150276 Supplier Invitrogen™ Supplier No. PA5150276
Only null left

Rabbit Polyclonal Antibody

NKRF Polyclonal Antibody for Western Blot, ICC/IF

This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predominantly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.

Specifications

Antigen NKRF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NKRF
Gene Accession No. O15226
Gene Alias 9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF
Gene Symbols NKRF
Host Species Rabbit
Immunogen Recombinant NFKB Repressing Factor (NKRF) (Met1-Val296).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less