Learn More
Invitrogen™ NKp30 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA5104376
Catalog No. PIPA5104376
Only null left
Explore available promotions
Description
Description
Antibody detects endogenous levels of total NCR3.
The protein encoded by this gene is a natural cytotoxicity receptor (NCR) that may aid NK cells in the lysis of tumor cells. The encoded protein interacts with CD3-zeta (CD247), a T-cell receptor. A single nucleotide polymorphism in the 5' untranslated region of this gene has been associated with mild malaria suceptibility. Three transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, May 2010].
Specifications
Specifications
NKp30
Polyclonal
Unconjugated
Ncr3
1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED
Rabbit
Affinity chromatography
RUO
259197, 294251
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14931, Q8CFD9
Ncr3
A synthesized peptide derived from human NCR3(Accession O14931), corresponding to amino acid residues A103-S153.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions
Product Suggestions