missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitron 98.0+%, TCI America™
$85.75 - $231.14
Chemical Identifiers
| CAS | 2218-94-2 |
|---|---|
| Molecular Formula | C20H16N4 |
| Molecular Weight (g/mol) | 312.376 |
| MDL Number | MFCD00005174 |
| InChI Key | CWGBFIRHYJNILV-UHFFFAOYSA-N |
| Synonym | nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt |
| PubChem CID | 720071 |
| IUPAC Name | (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide |
| SMILES | C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4 |
Chemical Identifiers
| 2218-94-2 | |
| 312.376 | |
| CWGBFIRHYJNILV-UHFFFAOYSA-N | |
| 720071 | |
| C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4 |
| C20H16N4 | |
| MFCD00005174 | |
| nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt | |
| (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide |
Specifications
| 2218-94-2 | |
| C20H16N4 | |
| 5 g | |
| CWGBFIRHYJNILV-UHFFFAOYSA-N | |
| (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide | |
| 720071 | |
| ≥98.0% (T) | |
| Nitron |
| 187°C | |
| MFCD00005174 | |
| nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt | |
| C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4 | |
| 312.376 | |
| 312.38 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes