accessibility menu, dialog, popup

UserName

NNNN

CAS RN 2218-94-2

IUPAC Name: 1,4-diphenyl-3-(phenylazanidyl)-1H-1,2,4λ⁵-triazol-4-ylium
Synonyms: 1,4-diphenyl-3-(phenylazanidyl)-1,2,4λ⁵-triazol-4-ylium
Molecular Weight (g/mol): 312.38
Molecular Formula: C20H16N4
InChi Key: CWGBFIRHYJNILV-UHFFFAOYSA-N
SMILES: [N-](C1=NN(C=[N+]1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)