missing translation for 'onlineSavingsMsg'
Learn More

Nitron, 95%, Thermo Scientific Chemicals

$45.18 - $101.15

Chemical Identifiers

CAS 2218-94-2
Molecular Formula C20H16N4
Molecular Weight (g/mol) 312.376
MDL Number MFCD00005174
InChI Key CWGBFIRHYJNILV-UHFFFAOYSA-N
Synonym nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
PubChem CID 720071
IUPAC Name (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide
SMILES C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1780903
View Documents
Thermo Scientific Chemicals
A1780903
1 g
Each for $45.18
Only null left
 
AAA1780906
View Documents
Thermo Scientific Chemicals
A1780906
5 g
Each for $101.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2218-94-2
312.376
CWGBFIRHYJNILV-UHFFFAOYSA-N
720071
C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4
C20H16N4
MFCD00005174
nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
(1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide

Specifications

2218-94-2
C20H16N4
1 g
nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4
312.376
312.38
Nitron
∼185°C (decomposition)
MFCD00005174
14,6613
CWGBFIRHYJNILV-UHFFFAOYSA-N
(1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide
720071
95%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 218-724-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only