missing translation for 'onlineSavingsMsg'
Learn More

Nitron, 95%, Thermo Scientific Chemicals

Catalog No. AAA1780903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1780903 1 g
AAA1780906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1780903 Supplier Thermo Scientific Chemicals Supplier No. A1780903
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2218-94-2
Molecular Formula C20H16N4
Molecular Weight (g/mol) 312.376
MDL Number MFCD00005174
InChI Key CWGBFIRHYJNILV-UHFFFAOYSA-N
Synonym nitron, unii-90im4z883z, 1,4-diphenyl-endoanilino-dihydrotriazole, 1,4-diphenyl-3-phenylamino-1h-1,2,4-triazolium inner salt, 1h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, hydroxide, inner salt, 1,4-diphenyl-3-phenylazamethylene-1,2,4-triazoline, 1,4-diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 4h-1,2,4-triazolium, 1,4-diphenyl-3-phenylamino-, inner salt, 4,5-dihydro-2,4-diphenyl-5-phenylimino-1h-1,2,4-triazolium inner salt
PubChem CID 720071
IUPAC Name (1,4-diphenyl-1,2,4-triazol-4-ium-3-yl)-phenylazanide
SMILES C1=CC=C(C=C1)[N-]C2=NN(C=[N+]2C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point ∼185°C (decomposition)
Quantity 1 g
Merck Index 14,6613
Formula Weight 312.38
Percent Purity 95%
Chemical Name or Material Nitron
EINECSNumber 218-724-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only