missing translation for 'onlineSavingsMsg'
Learn More

Nitromethane 98.0+%, TCI America™

Catalog No. p-7133677
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N020925G 25 g
N0209100G 100 g
N0209500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N020925G Supplier TCI America Supplier No. N020925G

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-52-5
Molecular Formula CH3NO2
Molecular Weight (g/mol) 61.04
MDL Number MFCD00007400
InChI Key LYGJENNIWJXYER-UHFFFAOYSA-N
Synonym methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4
PubChem CID 6375
ChEBI CHEBI:77701
IUPAC Name nitromethane
SMILES C[N+]([O-])=O

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 101°C
Quantity 25 g
UN Number 1261
Formula Weight 61.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Nitromethane
EINECSNumber (2)-0191
RTECSNumber PA9800000
TSCA Yes