missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitromethane 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -29°C |
| Color | Yellow |
| Boiling Point | 101°C |
| Quantity | 25 g |
| UN Number | 1261 |
| Formula Weight | 61.04 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Nitromethane |
Sécurité et manipulation
| EINECSNumber | (2)-0191 |
| RTECSNumber | PA9800000 |
| TSCA | Yes |