Learn More
Nitromethane, 96%, for HPLC
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Supplier: Thermo Scientific Chemicals 445670010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Nitromethane | |
| -29.0°C | |
| 100.0°C to 102.0°C | |
| 95% min. (GC) | |
| CH3NO2 | |
| CH3NO2 | |
| 1 L | |
| 15,6698 | |
| Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide | |
| C[N+]([O-])=O | |
| 61.04 | |
| 0.61 mPa.s (25°C) | |
| 61.04 | |
| HPLC |
| 75-52-5 | |
| 1.1200g/mL | |
| 35°C | |
| Glass bottle | |
| 1.3805 to 1.3825 | |
| MFCD00007400 | |
| 1.12 | |
| methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4 | |
| LYGJENNIWJXYER-UHFFFAOYSA-N | |
| nitromethane | |
| 6375 | |
| CHEBI:77701 | |
| 96% |
Chemical Identifiers
| 75-52-5 | |
| 61.04 | |
| LYGJENNIWJXYER-UHFFFAOYSA-N | |
| 6375 | |
| nitromethane |
| CH3NO2 | |
| MFCD00007400 | |
| methane, nitro, nitrocarbol, nitrometan, nitrometan polish, nitro-methane, unii-ru5wg8c3f4, ccris 1205, hsdb 106, meno2, ru5wg8c3f4 | |
| CHEBI:77701 | |
| C[N+]([O-])=O |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh
GHS Signal Word: Warning
EINECSNumber : 200-876-6
RUO – Research Use Only