missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin, 98%

CAS: 67-20-9 | C8H6N4O5 | 238.16 g/mol

$105.92 - $1112.14

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8H6N4O5
Molecular Weight (g/mol) 238.16
MDL Number MFCD00003224
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
PubChem CID 6604200
ChEBI CHEBI:71415
IUPAC Name 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2407914
View Documents
Thermo Scientific Chemicals
B2407914
25 g
Each for $105.92
Only null left
 
AAB2407922
View Documents
Thermo Scientific Chemicals
B2407922
100 g
Each for $250.67
Only null left
 
AAB2407936
View Documents
Thermo Scientific Chemicals
B2407936
500 g
Each for $1,112.14
Only null left
 
Description

Description

Nitrofurantoin is a nitrofuran antibiotic that is used as a substrate of bacterial nitrofuran reductase to study the interactions of active metabolites with DNA, ribosomal proteins and metabolic and pro-oxidant processes. It is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulfate and 5-nitro-2-furaldehyde diacetate. It shows activity against many Gram-positive and Gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corneybacteria, many strains of Escherichia coli.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrofurantoin is a nitrofuran antibiotic that is used as a substrate of bacterial nitrofuran reductase to study the interactions of active metabolites with DNA, ribosomal proteins and metabolic and pro-oxidant processes. It is an antibactericidal compound that has been historically prepared by the reaction of 1-aminohydantoin sulfate and 5-nitro-2-furaldehyde diacetate. It shows activity against many Gram-positive and Gram-negative bacteria. Nitrofurantoin is effective against enterococci, staphylococci, streptococci, corneybacteria, many strains of Escherichia coli.

Solubility
Soluble in water (<0.01g/100mL at 19°C), dimethyl sulfoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to light.
Specifications

Chemical Identifiers

67-20-9
238.16
NXFQHRVNIOXGAQ-YCRREMRBSA-N
6604200
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
C8H6N4O5
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
CHEBI:71415
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O

Specifications

67-20-9
C8H6N4O5
25 g
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
NXFQHRVNIOXGAQ-YCRREMRBSA-N
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
6604200
238.16
Nitrofurantoin
∼265°C (decomposition)
MFCD00003224
Hygroscopic; Light sensitive
Soluble in water (<0.01g/100 ml at 19°C),dimethyl sulfoxide and methanol.
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
238.16
CHEBI:71415
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H341-H351-H361-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.

P260-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P314-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H373

EINECSNumber : 200-646-5

RTECSNumber : MU2800000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only