missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin 98.0+%, TCI America™

$94.33 - $94.33

Chemical Identifiers

CAS 67-20-9
Molecular Formula C8H6N4O5
Molecular Weight (g/mol) 238.16
MDL Number MFCD00003224
InChI Key NXFQHRVNIOXGAQ-YCRREMRBSA-N
Synonym nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
PubChem CID 6604200
ChEBI CHEBI:71415
IUPAC Name 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
SMILES [O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N088325G
View Documents
TCI America
N088325G
25 g
Each for $94.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-20-9
238.16
NXFQHRVNIOXGAQ-YCRREMRBSA-N
6604200
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
C8H6N4O5
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
CHEBI:71415
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O

Specifications

67-20-9
Yellow
MFCD00003224
nitrofurantoin, macrodantin, 5-nitrofurantoin, furadantin, furadonine, nitrofurantoine, furadantine, furadantoin, furadoine, furadontin
[O-][N+](=O)C1=CC=C(O1)\C=N\N1CC(=O)NC1=O
238.16
CHEBI:71415
≥98.0% (HPLC,T)
Nitrofurantoin
268°C
C8H6N4O5
25 g
NXFQHRVNIOXGAQ-YCRREMRBSA-N
1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione
6604200
238.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0980

RTECSNumber : MU2800000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'