missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitrocellulose stripping buffer (10X)

Catalog No. AAJ62541AP
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62541AP 500 mL
AAJ62541AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62541AP Supplier Thermo Scientific Chemicals Supplier No. J62541AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for removal of secondary antibody after detection of highly abundant targets. Nitrocellulose Stripping Buffer may perform well with nitrocellulose membranes or antibody-dye conjugates. It is also employed for nucleic acid and protein related blotting works.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Product Type Western Blot Stripping Buffer
Form Liquid
Quantity 500 mL
Concentration 10X

RUO – Research Use Only