missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrocellulose Membranes, 0.45 μm

Catalog No. PI88025
Change view
Click to view available options
Dimensions (LxW):
10.5 x 7.9 cm
3.5 m x 30 cm
8 x 8 cm
12 x 8 cm
Format:
Roll
Sheet
Sufficient For:
≥120 Mini-gel Blots (When cut to 8 x 10 cm)
15 Mini-gel Blots
25 Mini-gel Blots
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (LxW) Format Sufficient For
PI88025 8 x 8 cm Sheet 15 Mini-gel Blots
PI88014 10.5 x 7.9 cm Sheet 15 Mini-gel Blots
PI77010 12 x 8 cm Sheet 25 Mini-gel Blots
PI88018 3.5 m x 30 cm Roll ≥120 Mini-gel Blots (When cut to 8 x 10 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI88025 Supplier Thermo Scientific™ Supplier No. 88025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods).

Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods (western blotting, dot-blot assays, and other protein or nucleic acid methods). The 0.45-μm pore size is well-suited for a wide range of western blotting conditions and ideal for the transfer of most proteins (>20 kDa) and nucleic acids (>300 bp).

Features include:
• 100% pure nitrocellulose membrane
• Convenient—save time with pre-cut sheets or choose the roll format for the flexibility to cut to the ideal size
• Compatible with commonly used transfer conditions and detection methods such as staining, immunodetection, chemiluminescence, fluorescence, and radiolabeling
• High-binding affinity—provides excellent protein binding, blocks easily, and does not require pre-wetting with alcohol
• Low background: produce high-quality data with low background signal in both chemiluminescent and fluorescent western blotting

Specifications

Length (Metric) 8 cm
Width (Metric) 8 cm
Description Nitrocellulose Membranes, 0.45 μm, 8 x 8 cm
Binding Capacity ∼45–60 μg/cm2 of BSA
Quantity 15 Pre-cut Membranes
Format Sheet
Material Nitrocellulose
Pore Size 0.45 μm
Content And Storage Store membranes flat at ambient temperature, away from chemical vapors. Some solvent vapors may partially dissolve the membranes, which will disrupt the pore structure. Keep membranes out of direct sunlight. Sunlight may cause the nitrocellulose to dry and become brittle.
Note: Nitrocellulose membrane filters are highly flammable. Keep away from heat and open flame. Flash point is approximately 2°C. Membranes can be sterilized by steam-autoclaving at 121°C.
Shipping Condition Room Temperature
Dimensions (LxW) 8 x 8 cm
Sufficient For 15 Mini-gel Blots
Show More Show Less
How can I store, strip, and reuse my western blot?

For nitrocellulose or PVDF membrane following Western blot detection using a chemiluminescent or fluorescent substrate system: Following transfer, air dry the membrane and place in an envelope, preferably on top of a supported surface to keep the membrane flat. The blot can be stored indefinitely at -80 degrees C. When ready to reprobe, prewet the PVDF blot with alcohol for a few seconds, followed by a few rinses with pure water to reduce the alcohol concentration. Then proceed as normal with blocking step.

FOR STRIPPING/REPROBING OF MEMBRANES: Harsh protocol (see NOTE below for modifications)

1) Submerge the membrane in stripping buffer (100 mM BME, 2% SDS, 62.5 mM Tris-HCl, pH 6.7) and incubate at 50 degrees C for 30 min with occasional agitation. If more stringent conditions necessary, incubate at 70 degrees C.

2) Wash 2 x 10 min in TBS-T/PBS-T at room temperature.

3) Block the membrane by immersing in 5% blocking reagent TBS-T or PBS-T for 1 hr at room temperature.

4) Immunodetection

NOTE: Often you don't need such harsh conditions to remove antibodies from their proteins. The stringency of one or several of the variables can be decreased: lower the temperature, decrease the time, less BME, less SDS, etc. An especially mild but still often effective stripping protocol is lower pH incubation. Example: pH 2.0 Tris 50-100 mM, 30-60 min incubation (you may do two incubations if you wish). Then rinse and block as usual. If you do not wish to re-use the membrane immediately after stripping, you can store the membrane in plastic wrap (wet, you do not want it to dry out). Another simple, mild stripping buffer is 0.1 M glycine•HCl (pH 2.5-3.0), incubation 30 min to 2 hrs room temperature or 37 degrees C, depending on the antibody.

Are your PVDF and nitrocellulose membranes compatible with the Li-COR instrument?

Yes, both our PVDF and nitrocellulose membranes are compatible with the Li-COR instrument.