missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrobenzene, Reagents
Nitrobenzene, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2257500500C
Specifications
| Nitrobenzene | |
| 211°C | |
| Pale Yellow | |
| 5.6°C to 5.8°C | |
| C6H5NO2 | |
| 1.2037 | |
| LQNUZADURLCDLV-UHFFFAOYSA-N | |
| nitrobenzene | |
| Reagent |
| 98-95-3 | |
| 5.6°C to 5.8°C | |
| 211°C | |
| Glass Bottle | |
| 500 mL | |
| Nitrobenzol, Oil of Mirbane | |
| [O-][N+](=O)C1=CC=CC=C1 | |
| 123.11 | |
| Liquid |
Chemical Identifiers
| 98-95-3 | |
| 123.11 | |
| Nitrobenzol, Oil of Mirbane | |
| [O-][N+](=O)C1=CC=CC=C1 |
| C6H5NO2 | |
| LQNUZADURLCDLV-UHFFFAOYSA-N | |
| nitrobenzene |