missing translation for 'onlineSavingsMsg'
Learn More

Nitrobenzene, Reagents

Catalog No. p-200020877
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337153 4 L
01337152 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337153 Supplier Labchem Inc Supplier No. C22575004C
Only null left

Nitrobenzene, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 98-95-3
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
InChI Key LQNUZADURLCDLV-UHFFFAOYSA-N
Synonym Nitrobenzol, Oil of Mirbane
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1

Specifications

Boiling Range 211°C
Melting Point 5.6°C to 5.8°C
Color Pale Yellow
Boiling Point 211°C
Melting Range 5.6°C to 5.8°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 1.2037
Grade Reagent
Physical Form Liquid
Chemical Name or Material Nitrobenzene
Show More Show Less