missing translation for 'onlineSavingsMsg'
Learn More

nitro-Grela 97.0+%, TCI America™

Catalog No. N1060100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1060100MG 100 mg
N1060500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1060100MG Supplier TCI America Supplier No. N1060100MG
Only null left

Chemical Identifiers

CAS 502964-52-5
Molecular Formula C31H37Cl2N3O3Ru
MDL Number MFCD28411645
Synonym [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro[(2-isopropoxy-5-nitrobenzylidene)]ruthenium(VI)

Specifications

Quantity 100 mg
Formula Weight 671.63
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material nitro-Grela