missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride monohydrate, 90%

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.644 g/mol

$370.93 - $370.93

Specifications

Chemical Name or Material Nitro Blue Tetrazolium Chloride monohydrate
CAS 298-83-9 , 67-56-1 , 7732-18-5
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC128590010
View Documents
Thermo Scientific Chemicals
128590010
1 g
Each for $370.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nitro Blue Tetrazolium Chloride monohydrate
88.0
90%
MFCD00150013
FSVCQIDHPKZJSO-UHFFFAOYSA-L
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
817.644
CHEBI:9505
10% max. (105°C)
Glass bottle
Powder
298-83-9 , 67-56-1 , 7732-18-5
100.0
C40H30Cl2N10O6
Nitro BT, Nitro blue tetrazolium
Authentic
2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
9281
867.69
≥88%
Yellow
Videos
SDS
Documents

Documents

RUO – Research Use Only