missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nitro Blue Tetrazolium Chloride, 90%

CAS: 298-83-9 | C40H30Cl2N10O6 | 817.65 g/mol

$392.94 - $392.94

Chemical Identifiers

CAS 298-83-9
Molecular Formula C40H30Cl2N10O6
Molecular Weight (g/mol) 817.65
InChI Key FSVCQIDHPKZJSO-UHFFFAOYSA-L
Synonym nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
PubChem CID 9281
ChEBI CHEBI:9505
IUPAC Name 2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
SMILES COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC415920010
SDS View Documents
Thermo Scientific Chemicals
415920010
1 g Glass bottle
Each for $392.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

298-83-9
817.65
nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
CHEBI:9505
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
C40H30Cl2N10O6
FSVCQIDHPKZJSO-UHFFFAOYSA-L
9281
2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride

Specifications

298-83-9
88% min. (Argentometry)
C40H30Cl2N10O6
nitrotetrazolium blue chloride, nitro bt, nitro blue tetrazolium, nbt, nitro blue tetrazolium chloride, p-nitrotetrazolium blue, nitrotetrazolium blue, nitroblue tetrazolium chloride, p-nitro blue tetrazolium chloride, nitro tetrazolium bt
FSVCQIDHPKZJSO-UHFFFAOYSA-L
2-[2-methoxy-4-[3-methoxy-4-[3-(4-nitrophenyl)-5-phenyltetrazol-2-ium-2-yl]phenyl]phenyl]-3-(4-nitrophenyl)-5-phenyltetrazol-2-ium;dichloride
9281
817.65
Nitro Blue Tetrazolium Chloride
189°C
Glass bottle
1 g
Solubility in water: soluble. Other solubilities: soluble in methanol
COC1=C(C=CC(=C1)C2=CC(=C(C=C2)[N+]3=NC(=NN3C4=CC=C(C=C4)[N+](=O)[O-])C5=CC=CC=C5)OC)[N+]6=NC(=NN6C7=CC=C(C=C7)[N+](=O)[O-])C8=CC=CC=C8.[Cl-].[Cl-]
817.65
CHEBI:9505
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
IF IN EYES: Rinse cautiously with water f

GHS Signal Word: Warning

EINECSNumber : 206-067-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only