Learn More
Ansell™ BioClean-D™ Sterile Coveralls with Collar and Thumb Loop
Disposable sterile coverall with thumb loop for secure hold.
Supplier:  Ansell™ SBDCCT2XL
 View more versions of this product
Catalog No. 19-668-337
Only null left
Explore available promotions
Description
Description
  • Low linting, anti-static CleanTough™ Fabric made
  • Sterilized by Gamma irradiation
  • Anti-Static properties
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Zip front with flap cover
  • Elasticated back, cuffs and ankles
  • Available in sizes ranging from S through 4XL
  • ASTM D6978 05 Tested
  • HELMKE DRUM Tested
  • Low-particulate
  • Sterility Assurance Level 10-6
Compliance
USP 797, USP 800, 503B
Specifications
Specifications
Cleanroom Coveralls
White
ISO 9001:2015
S-BDCCT
116 to 124 cm
6 ft. 2 in. to 6 ft. 4 in.
2X-Large
Sterile
EN ISO 13982-1:2004+A1:2010, EN ISO 13034:2005+A1:2009, EN ISO 14325, ASTM F739-12, EN 1149-5:2008, EN530, EN 7854, EN 6530, EN 13934-1, EN9073-4, EN 863 and EN 13935-2, ISO 11137-1:2006
45-48 in.
Cleanroom, Pharma, Controlled Environments, PPE, 503B Facilities, Manufacturing, Industrial
188 to 194 cm
Product Suggestions
Product Suggestions