Learn More
StatLab™ Nitrical Decalcifier
A safe and gentle standard AFIP formulation.
$72.96
Specifications
| Solution Type | Decalcifier |
|---|
Alert:
Harmful if swallowed, in contact with skin, or inhaled.
Description
- Made from semiconductor grade acid and highly purified reagent grade water
- Very mild solution that works rapidly with great results
- Can be used for all types of specimens from bone marrow to cortical bone
Specifications
| Decalcifier |