Learn More
StatLab™ Nitrical Decalcifier
Catalog No. STL121332
Change view
Click to view available options
Quantity:
4 x 32 oz. Bottle
Catalog No. Quantity
STL121332 4 x 32 oz. Bottle
1 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL121332 Supplier StatLab™ Supplier No. 121332
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or inhaled.
A safe and gentle standard AFIP formulation.
  • Made from semiconductor grade acid and highly purified reagent grade water
  • Very mild solution that works rapidly with great results
  • Can be used for all types of specimens from bone marrow to cortical bone
Specifications
Quantity 4 x 32 oz. Bottle
Solution Type Decalcifier