missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 67-69%, Honeywell Fluka™

Catalog No. 60044473
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 L
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044473 250 mL Poly Bottle
6001110 500 mL Poly Bottle
60044474 2 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60044473 Supplier Honeywell Chemicals Supplier No. 02650250ML
Only null left

TraceSELECT™ Ultra, for ultratrace analysis, 67-69%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Boiling Point 122°C
Density 1.420 g/cm3
Packaging Poly Bottle
Vapor Pressure 8mmHg (20°C)
Assay Percent Range 64.5 to 70%
Quantity 250 mL
Linear Formula HNO3
UN Number UN2031
Formula Weight 63.01
Percent Purity 67 to 69%
Grade Ultratrace
Chemical Name or Material Nitric acid, TRACESELECT™ Ultra, for UltraTrace Analysis, 65-71%,
Show More Show Less
Hazard Statement P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
Precautionary Statement H272-H290-H314
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Nitric acid
Recommended Storage Room Temp