missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (TraceMetal™ Grade), Fisher Chemical™

Catalog No. A509P500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A509P500 500 mL HDPE Bottle
A509P212 2.5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A509P500 Supplier Seastar Chemicals Inc Supplier No. S010101TFIQ03
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Nitric Acid (TraceMetal™ Grade)

  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured,purified, and packaged in a Class 1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Two year shelf life from time of manufacture - available on COA. 45 mm bottle neck size

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Packaging HDPE Bottle
Vapor Pressure 0.94 kPa at 20°C
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 500 mL
Physical Form Liquid
UN Number 2031
Formula Weight 63.01
Percent Purity 67 to 70% (HNO3, w/w)
Grade TraceMetal
Chemical Name or Material Nitric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8: Corrosive