missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, Technical, Spectrum™ Chemical
N1088, 7697-37-2, HNO3
$486.07 - $486.07
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Description
Spectrum™ Chemical Nitric Acid, Technical is a strong oxidizing agent and a highly corrosive mineral acid. Primarily used as a reagent, the pure compound is colorless and used for nitration, typically to an organic molecule.
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| nitric acid |
Specifications
| 7697-37-2,7732-18-5 | |
| Glass Bottle | |
| HNO3 | |
| UN2031 | |
| O[N+]([O-])=O | |
| 63.01 | |
| Nitric Acid |
| 70%,30% | |
| 2.5 L | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| Technical |