missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone, ≥65%, Honeywell Fluka™
Puriss. p.a., Reag. ISO, Reag. Ph. Eur., for determinations with dithizone,≥65%
$250.95 - $250.95
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Specifications
| 7697-37-2 | |
| Plastic Bottle | |
| 8mmHg (20°C) | |
| HNO3 | |
| 2031 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 63.01g/mol | |
| puriss. p.a. |
| 1.37 to 1.41g/mL at 20°C | |
| ≤0.0005% (as SO4) | |
| 1 L | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| ≥65% | |
| Nitric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P210-P220-P260-P280-P305 + P351 + P338-P370 + P378
H272-H290-H314