missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric acid, Puriss. p.a., ACS Reagent, Reag. ISO, ≥69%, Honeywell Fluka
Puriss. p.a., ACS Reagent, Reag. ISO,≥69%
$224.13 - $224.13
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Specifications
| 7697-37-2,7732-18-5 | |
| ≥50.0000%,≥30.0000% | |
| <70.0000%,<50.0000% | |
| 2.5 L | |
| HNO3 | |
| UN2031 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 63.01 | |
| ACS Reagent/Reagent ISO |
| 122°C | |
| 1.413Â g/mLÂ at 20Â °C(lit.) | |
| Glass Bottle | |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| >69% | |
| Nitric acid, Puriss. p.a., ACS Reagent, Reag. ISO, ≥69% |
Safety and Handling
ShelfLife : 720 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Nitric acid
Recommended Storage : Room Temp