missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, puriss., Honeywell Fluka™

Puriss., 64-66%

$314.32 - $314.32

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001102
View Documents
Honeywell-Fluka
0700625L
2.5 L
Each for $314.32
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Specifications

7697-37-2
1.37 to 1.41g/mL at 20°C
8mmHg (20°C)
HNO3
2031
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01g/mol
Nitric Acid
64 to 66%
≤0.0005% (as SO4)
2.5 L
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
puriss.
Videos
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H314

SDS
Documents

Documents