missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, GR, ACS, MilliporeSigma™

Catalog No. 361042005
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glas bottle (Pre packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361042005 500 mL Glas bottle (Pre packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361042005 Supplier Technidata Supplier No. NX0409P5

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH <1 (H2O, 20°C)
Appearance To pass test
Melting Point -32°C
Boiling Point 121°C (1013hPa)
CAS Min % Nitric acid: 50% to 70%
Odor Stinging
Packaging Glas bottle (Pre packed)
Residue after Ignition 5ppm
Vapor Pressure 9.4 hPa (20°C)
Quantity 500 mL
Physical Form Liquid
Percent Purity 68.0 to 70.0%
Chemical Name or Material Nitric Acid
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category H272 May intensify fire; oxidizer. H290 May be corrosive to metals. H314 Causes severe skin burns and eye damage
Hazard Statement P280 Wear protective gloves/ protective clothing/ eye protection/ face protection. P301 + P330 + P331 IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. P305 + P351 + P338 IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. P308 + P310 IF exposed or concerned: immediately call a POISON CENTER or doctor/ physician
DOTInformation 5.1B Oxidizing hazardous materials