missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid concentrate, For 1L standard solution, 0.1 M HNO3 (0.1N), Honeywell Fluka™

For 1L standard solution, 0.1 M HNO3 (0.1N)

$225.13 - $225.13

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6001099
View Documents
Honeywell Chemicals
320421EA
1 Ea. Ampoule
Each for $225.13
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O

Specifications

7697-37-2
1 Ea.
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
O[N+]([O-])=O
63.01
CHEBI:48107
Nitric Acid Concentrate
Ampoule
HNO3
2031
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01g/mol
Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

H290-H314

EINECSNumber : 231-714-2

SDS
Documents

Documents