missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid concentrate, For 1L standard solution, 1.0 M HNO3 (1.0N), Solstice

Catalog No. 6001100
Change view
Click to view available options
Quantity:
1 Pc.
Packaging:
Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001100 1 Pc. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6001100 Supplier Solstice Supplier No. 382741EA

For 1L standard solution, 1.0 M HNO3 (1.0N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Packaging Ampoule
Quantity 1 Pc.
UN Number UN2031B
Formula Weight 63.01g/mol
Chemical Name or Material Nitric acid concentrate
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338