Learn More
Nitric Acid (Certified ACS Plus), Fisher Chemical™
Nitric Acid material may darken during storage due to photochemical reaction
Supplier: Thermo Fisher Scientific A200612GAL
Specifications
| -1.0 | |
| Colorless Yellow | |
| 7697-37-2,7732-18-5 | |
| 1.40g/cm³ | |
| Pass Test | |
| 2ppm max. | |
| about 15.8 | |
| 6.5 gal. | |
| Liquid | |
| 2031 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 63.01 | |
| Certified ACS Plus |
| Nitric Acid | |
| -41°C | |
| 120.5°C | |
| 0.2ppm max. | |
| Glass Carboy | |
| 0.94 kPa at 20°C | |
| 70% | |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| 68.0 to 70.0% (w/w) |
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Safety and Handling
DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:4
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive