missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, Certified, 50% (v/v) ±1%, LabChem™
Supplier: LabChem LC177703
Specifications
| Nitric Acid (1+1) | |
| 7697-37-2,7732-18-5 | |
| 60,40 | |
| HDPE Bottle | |
| Traceable to NIST | |
| 2.5 L | |
| Liquid | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 63.01 | |
| Certified |
| Colorless | |
| 1.25g/mL | |
| Passes Test | |
| 1.25g/mL | |
| Nitrogen oxides; Thermal decomposition generates: Corrosive vapors | |
| HNO3 | |
| HNO3 | |
| UN2031 | |
| Soluble in water | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| 50% ±1% (v/v) |
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse m
Danger
Recommended Storage : Room Temperature