missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, Certified, 2.00N ±0.02N, LabChem™

Supplier:  LabChem LC178502

 View more versions of this product

Catalog No. LC178502


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nitric Acid
7697-37-2,7732-18-5
1.07g/mL
Passes Test
1.07g/mL
Nitrogen oxides; Thermal decomposition generates: Corrosive vapors
HNO3
HNO3
UN2031
Soluble in water
O[N+]([O-])=O
63.01
CHEBI:48107
Certified
Colorless
2.00N ±0.02N
88,12
Amber Glass
Traceable to NIST
1 L
Liquid
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions