Learn More
Nitric Acid, Certified, 1.000N ±0.005N, LabChem™
Supplier: LabChem LC178402
Specifications
| Nitric Acid | |
| 7697-37-2,7732-18-5 | |
| 1.03g/mL | |
| Passes Test | |
| 1.03g/mL | |
| Nitrogen oxides; Thermal decomposition generates: Corrosive vapors | |
| HNO3 | |
| HNO3 | |
| UN2031 | |
| Soluble in water | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Certified |
| Colorless | |
| 1.000N ±0.005N | |
| 93.9,6.1 | |
| Amber Glass | |
| Traceable to NIST | |
| 1 L | |
| Liquid | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 63.01 |
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature