missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, ACS Grade, 68.0% to 70.0% (w/w), LabChem™

Supplier:  LabChem LC177003

 View more versions of this product

Catalog No. LC177003

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nitric Acid
7697-37-2
Passes Test
Nitrogen oxides; oxygen
HNO3
HNO3
2031
Soluble in water
O[N+]([O-])=O
63.01
CHEBI:48107
68.0 to 70.0% (w/w)
Colorless
100
HDPE Bottle
2.5 L
Liquid
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
63.01
ACS

Chemical Identifiers

7697-37-2
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011349
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
O[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Keep only in original container.
Do no

Danger

DOTInformation : DOT Class 8: Corrosive

EINECSNumber : 231-714-2

Recommended Storage : Room Temperature

SDS Documents

Documents

Promotions

Promotions