missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, ACS, 68.0-70.0%, Thermo Scientific Chemicals

Catalog No. AA33260P2
Change view
Click to view available options
Quantity:
1 lb.
7 lb.
6 x 7 lb.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33260P2 1 lb.
AA33260P7 7 lb.
AA33260R1 6 x 7 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA33260P2 Supplier Thermo Scientific Chemicals Supplier No. 33260P2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Main commercial usage of Nitric acid, ACS, 68.0-70.0% is in the production of fertilizers and reagent for nitration. It is used in the applications for the production of explosives, nylon precursors and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors and in purification processes for raw carbon nanotubes, artificially age pine and maple.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Boiling Point 121°C
Density 1.41
Assay Percent Range 68.0-70.0%
Quantity 1 lb.
Physical Form Liquid
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Solubility Information Miscible with water and ether.
Formula Weight 63.01
Grade ACS
Chemical Name or Material Nitric Acid
Show More Show Less
Hazard Category H272-H290-H314-H331
Precautionary Statement P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 231-714-2
RTECSNumber QU5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only