missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 70.0%, MilliporeSigma™

Catalog No. 50947889
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947889 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947889 Supplier Technidata Supplier No. NX04074

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH < 1 (H2O, 20°C)
Melting Point −32°C
Boiling Point 121°C (1013 hPa)
Packaging Plastic Bottle
Vapor Pressure 9.4 hPa (20°C)
Assay Percent Range 67 to 70%
Quantity 2.5 L
Recommended Storage Store at room temperature