missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 69.5% LM Prime, Transene

Catalog No. 502744712
Encompass
Catalog No. 502744712 Supplier Doe and Ingalls Management LLC Supplier No. T0701499GAL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specially Produced For The Electronic Industry

Nitric Acid 69.5% semiconductor grade is used in manufacturing processes serving as a chemical bath for cleaning silicon wafers. One of its properties is the elimination of particles and contaminants from the semiconductor surfaces after the etching process. In addition, it enhances the surface smoothness, flatness and removes the oxide layer from the surfaces of semiconductors.

  • Common Uses and Applications:
  • Cleaning silicon wafers
  • Solar energy
  • Metal processing
  • LCD panels
  • RAM devices
  • Industries
  • Electronics/Semiconductor
  • Industrial
  • R and D Laboratory

Safety and Shipping Information: DOT: Nitric acid, solution, 8, UN2031, PG II, Hazmat Fee Required.

Chemical Identifiers

CAS 7697-37-2
Molecular Weight (g/mol) 63 g/mol

Specifications

Packaging HDPE
Quantity 1 gal.
Percent Purity 69.5%
Grade Electronic
Chemical Name or Material Nitric Acid