missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 67-70%, OmniTrace Ultra, MilliporeSigma™

Catalog No. MNX04086
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNX04086 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNX04086 Supplier Technidata Supplier No. NX04086

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Assay Percent Range 67 to 70%
Quantity 1 L
Physical Form Liquid
Linear Formula HNO3
Formula Weight 63.1
Chemical Name or Material Nitric Acid, OmniTrace™ Ultra
Recommended Storage Room Temperature