missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid (65% w/w/Certified), Fisher Chemical

Catalog No. A206C212
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A206C212 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A206C212 Supplier Thermo Fisher Scientific Supplier No. FLA206C212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH 1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Heavy Metals (as Pb) 0.1ppm max.
Packaging Glass Bottle
Residue after Ignition 2ppm max.
Vapor Pressure 0.94 kPa at 20°C
Quantity 2.5 L
Physical Form Liquid
UN Number 2031
Formula Weight 63.01
Percent Purity 64.8 to 65.2%
Grade Certified
Concentration or Composition (by Analyte or Components) 65% w/w
Chemical Name or Material Nitric Acid, 65% w/w
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive