missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 65%, Suprapur™, MilliporeSigma™
$362.79 - $362.79
Chemical Identifiers
| CAS | 7697-37-2 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
M1004410250
|
Technidata
1004410250 |
250 mL | Glass Bottle |
Each for $362.79
|
|
||||
Chemical Identifiers
| 7697-37-2 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Specifications
| Colorless | |
| 1.39g/cm3 (20°C) | |
| 9.4 hPa (20°C) | |
| 250 mL | |
| Liquid | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| Suprapur |
| 7697-37-2 | |
| Glass Bottle | |
| 65% | |
| HNO3 | |
| MFCD00011349 | |
| Soluble (20°C) | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Nitric Acid, Suprapur™ |
Safety and Handling
SignalWord: Danger
Recommended Storage : Store below +25°C