missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 65%, Suprapur™, MilliporeSigma™

Catalog No. M1004410250
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1004410250 250 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1004410250 Supplier Technidata Supplier No. 1.00441.0250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless
Density 1.39g/cm3 (20°C)
Packaging Glass Bottle
Vapor Pressure 9.4 hPa (20°C)
Assay Percent Range 65%
Quantity 250 mL
Physical Form Liquid
Solubility Information Soluble (20°C)
Grade Suprapur
Chemical Name or Material Nitric Acid, Suprapur™
Hazard Category SignalWord: Danger
Recommended Storage Store below +25°C