missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 20% (w/v) Aqueous Solution, Spectrum™ Chemical

$307.79 - $362.00

Chemical Identifiers

CAS 7697-37-2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611995
View Documents
Spectrum Chemical Mfg Cor
N1701LT
1 L Amber Glass Bottle
Each for $307.79
Only null left
 
18611996
View Documents
Spectrum Chemical Mfg Cor
N17025LTPV
2.5 L Poly-Coated Glass Bottle
Each for $362.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7697-37-2

Specifications

7697-37-2,7732-18-5
1 L
Ungraded
Amber Glass Bottle
20% ±0.5%
Nitric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.