missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 10% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 53201
Specifications
| Nitric Acid | |
| 7732-18-5,7697-37-2 | |
| 10% (v/v) | |
| Natural Poly Bottle | |
| 4 L | |
| Liquid | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Laboratory |
| Colorless to Yellow | |
| 89.0,9.0 | |
| 91.0,11.0 | |
| Aqueous Solution | |
| HNO3 | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 10% V/V |
Chemical Identifiers
| 7732-18-5 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |